The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020967.2 | NP_066018.1 | 4 - 5 | 651 | 58 |
XM_005260474.3 | XP_005260531.1 | 3 - 4 | 605 | 58 | |
XM_011528951.2 | XP_011527253.1 | 4 - 5 | 666 | 58 | |
XM_017027988.1 | XP_016883477.1 | 3 - 4 | 605 | 58 | |
GenBank mRNA | AB046857.1 | - | 2 - 3 | 199 | 58 |
AF230533.1 | - | 4 - 5 | 623 | 58 | |
AF470686.1 | - | 3 - 4 | 1027 | 58 | |
BC140836.1 | - | 3 - 4 | 540 | 58 | |
BC151133.1 | - | 3 - 4 | 540 | 58 |