Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCB1
Entrez Gene ID
23236
Gene Name
phospholipase C beta 1
Gene Aliases
EIEE12, PI-PLC, PLC-154, PLC-I, PLC-beta-1, PLC154, PLCB1A, PLCB1B
Chromosome Location
Chr.20: 8132236 - 8884911 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015192.3 NP_056007.1 4 - 5 768 81
NM_182734.2 NP_877398.1 4 - 5 768 81
XM_011529199.2 XP_011527501.1 4 - 5 728 81
XM_011529200.2 XP_011527502.1 2 - 3 392 81
XM_011529201.2 XP_011527503.1 2 - 3 202 81
XM_017027752.1 XP_016883241.1 4 - 5 728 81
XM_017027753.1 XP_016883242.1 4 - 5 355 81
GenBank mRNA AB011153.3 - 4 - 5 768 81
AJ278313.1 - 4 - 5 384 81
AJ278314.1 - 4 - 5 384 81
AK127693.1 - 2 - 3 183 81
AK299195.1 - 4 - 5 403 81
AK310597.1 - 5 - 6 696 81
AY004175.1 - 4 - 5 366 81
BC069420.1 - 4 - 5 384 81
BC117231.1 - 4 - 5 439 81

View Full Product Details