The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171971.2 | NP_001165442.1 | 8 - 9 | 907 | 63 |
NM_033100.3 | NP_149091.1 | 8 - 9 | 907 | 63 | |
XM_011540337.1 | XP_011538639.1 | 7 - 8 | 1398 | 63 | |
XM_011540338.1 | XP_011538640.1 | 7 - 8 | 1398 | 63 | |
XM_011540340.2 | XP_011538642.1 | 7 - 8 | 1398 | 63 | |
GenBank mRNA | AB053448.1 | - | 8 - 9 | 780 | 63 |
AB593154.1 | - | 8 - 9 | 907 | 63 | |
AB593155.1 | - | 8 - 9 | 893 | 63 | |
AK124752.1 | - | 2 - 3 | 161 | 63 | |
BC038799.1 | - | 8 - 9 | 897 | 63 |