The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000834.3 | NP_000825.2 | 3 - 4 | 1189 | 79 |
XM_011520628.2 | XP_011518930.1 | 4 - 5 | 1982 | 79 | |
XM_011520629.2 | XP_011518931.1 | 5 - 6 | 1789 | 79 | |
XM_017019219.1 | XP_016874708.1 | 4 - 5 | 1537 | 79 | |
GenBank mRNA | BC113618.1 | - | 3 - 4 | 1088 | 79 |
BC113620.1 | - | 3 - 4 | 1088 | 79 | |
U11287.1 | - | 3 - 4 | 1220 | 79 | |
U88963.1 | - | 3 - 4 | 1189 | 79 | |
U90278.1 | - | 3 - 4 | 1219 | 79 |