The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031488.4 | NP_113676.2 | 9 - 10 | 1230 | 52 |
XM_011530420.2 | XP_011528722.1 | 8 - 9 | 1213 | 52 | |
XM_011530421.2 | XP_011528723.1 | 9 - 10 | 1299 | 52 | |
XM_017028976.1 | XP_016884465.1 | 7 - 8 | 1007 | 52 | |
XM_017028977.1 | XP_016884466.1 | 8 - 9 | 1526 | 52 | |
XM_017028978.1 | XP_016884467.1 | 9 - 10 | 1481 | 52 | |
XM_017028979.1 | XP_016884468.1 | 9 - 10 | 1426 | 52 | |
XM_017028980.1 | XP_016884469.1 | 8 - 9 | 1113 | 52 | |
GenBank mRNA | AJ305226.1 | - | 9 - 10 | 1215 | 52 |
AJ305227.1 | - | 9 - 10 | 1215 | 52 | |
AK056443.1 | - | 9 - 10 | 1227 | 52 | |
AK074091.1 | - | 8 - 9 | 3434 | 52 | |
AK097052.1 | - | 9 - 10 | 1299 | 52 | |
AK307763.1 | - | 9 - 10 | 1222 | 52 | |
AL136564.1 | - | 9 - 10 | 1230 | 52 | |
BC017191.2 | - | 9 - 10 | 1219 | 52 | |
CR456482.1 | - | 9 - 10 | 1197 | 52 | |
KU178799.1 | - | 9 - 10 | 1176 | 52 | |
KU178800.1 | - | 9 - 10 | 1176 | 52 |