The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318189.1 | NP_001305118.1 | 11 - 12 | 1269 | 90 |
NM_144587.3 | NP_653188.2 | 11 - 12 | 1266 | 90 | |
XM_011539239.2 | XP_011537541.1 | 10 - 11 | 1700 | 90 | |
XM_011539240.2 | XP_011537542.1 | 10 - 11 | 1675 | 90 | |
XM_011539241.2 | XP_011537543.1 | 9 - 10 | 1618 | 90 | |
XM_011539242.2 | XP_011537544.1 | 8 - 9 | 1499 | 90 | |
XM_017015636.1 | XP_016871125.1 | 10 - 11 | 2349 | 90 | |
XM_017015637.1 | XP_016871126.1 | 7 - 8 | 1974 | 90 | |
GenBank mRNA | AK058088.1 | - | 11 - 12 | 1256 | 90 |
BC109261.1 | - | 11 - 12 | 1240 | 90 | |
BC109262.1 | - | 11 - 12 | 1240 | 90 |