The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014023.3 | NP_054742.2 | 2 - 3 | 302 | 60 |
GenBank mRNA | AB023199.1 | - | 2 - 3 | 273 | 60 |
AK292591.1 | - | 2 - 3 | 265 | 60 | |
AL136827.1 | - | 2 - 3 | 280 | 60 | |
BC018044.1 | - | 2 - 3 | 331 | 60 |