The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004948.3 | NP_004939.1 | 15 - 16 | 2746 | 109 |
GenBank mRNA | X72925.1 | - | 15 - 16 | 2746 | 109 |
Z34522.1 | - | 15 - 16 | 2674 | 109 |