Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location
Chr.12: 30709552 - 30754994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002259.2 NP_001002259.1 9 - 10 2655 118
NM_001206856.2 NP_001193785.1 9 - 10 2655 118
NM_001319842.1 NP_001306771.1 8 - 9 2572 118
NM_001319843.1 NP_001306772.1 9 - 10 2655 118
NM_001319844.1 NP_001306773.1 10 - 11 1910 118
NM_001319845.1 NP_001306774.1 10 - 11 1910 118
NM_001319846.1 NP_001306775.1 10 - 11 1900 118
NM_023925.4 NP_076414.2 9 - 10 2655 118
NM_032156.4 NP_115532.3 9 - 10 2655 118
XM_006719138.3 XP_006719201.1 9 - 10 2778 118
XM_006719139.3 XP_006719202.1 9 - 10 2778 118
XM_006719140.3 XP_006719203.1 9 - 10 2778 118
XM_006719142.3 XP_006719205.1 9 - 10 2778 118
XM_006719144.3 XP_006719207.1 9 - 10 2778 118
XM_006719145.3 XP_006719208.1 9 - 10 2778 118
XM_006719146.3 XP_006719209.1 9 - 10 2778 118
XM_006719147.3 XP_006719210.1 9 - 10 2778 118
XM_017019851.1 XP_016875340.1 9 - 10 2778 118
XM_017019852.1 XP_016875341.1 10 - 11 1873 118
XM_017019853.1 XP_016875342.1 10 - 11 1943 118
XM_017019854.1 XP_016875343.1 9 - 10 2778 118
XM_017019855.1 XP_016875344.1 10 - 11 1900 118
XM_017019856.1 XP_016875345.1 10 - 11 1900 118
XM_017019857.1 XP_016875346.1 10 - 11 1873 118
XM_017019858.1 XP_016875347.1 10 - 11 1900 118
XM_017019859.1 XP_016875348.1 10 - 11 1873 118
XM_017019860.1 XP_016875349.1 9 - 10 2778 118
XM_017019861.1 XP_016875350.1 10 - 11 1900 118
XM_017019862.1 XP_016875351.1 9 - 10 2778 118
XM_017019863.1 XP_016875352.1 9 - 10 2778 118
XM_017019864.1 XP_016875353.1 9 - 10 2778 118
XM_017019865.1 XP_016875354.1 9 - 10 2778 118
XM_017019866.1 XP_016875355.1 10 - 11 1873 118
XM_017019867.1 XP_016875356.1 10 - 11 1900 118
XM_017019868.1 XP_016875357.1 10 - 11 1873 118
XM_017019869.1 XP_016875358.1 10 - 11 1900 118
XM_017019870.1 XP_016875359.1 10 - 11 1873 118
XM_017019871.1 XP_016875360.1 10 - 11 1944 118
XM_017019872.1 XP_016875361.1 10 - 11 1900 118
XM_017019873.1 XP_016875362.1 10 - 11 1900 118
XM_017019874.1 XP_016875363.1 9 - 10 1790 118
XM_017019875.1 XP_016875364.1 10 - 11 1900 118
XM_017019876.1 XP_016875365.1 7 - 8 1401 118
XM_017019877.1 XP_016875366.1 7 - 8 1401 118
XM_017019878.1 XP_016875367.1 7 - 8 1401 118
XM_017019879.1 XP_016875368.1 9 - 10 1790 118
XM_017019880.1 XP_016875369.1 7 - 8 1401 118
XM_017019881.1 XP_016875370.1 9 - 10 1790 118
XM_017019882.1 XP_016875371.1 9 - 10 1790 118
XM_017019883.1 XP_016875372.1 7 - 8 1401 118
GenBank mRNA AB058776.1 - - 950 118
AF525416.1 - 1 - 2 166 118
AK021453.1 - 8 - 9 1422 118
AL834212.1 - 6 - 7 1142 118
AY074490.1 - 9 - 10 2655 118
AY074491.1 - 8 - 9 2572 118
BC117672.1 - 9 - 10 2452 118
BC117673.1 - 9 - 10 2452 118

View Full Product Details