The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039960.2 | NP_001035049.1 | 7 - 8 | 1029 | 122 |
NM_001258401.2 | NP_001245330.1 | 7 - 8 | 975 | 122 | |
NM_001258402.1 | NP_001245331.1 | 7 - 8 | 1029 | 122 | |
NM_001258403.1 | NP_001245332.1 | 7 - 8 | 1101 | 122 | |
NM_001267615.1 | NP_001254544.1 | 7 - 8 | 975 | 122 | |
XM_006719700.1 | XP_006719763.1 | 7 - 8 | 1030 | 122 | |
XM_011539010.1 | XP_011537312.1 | 7 - 8 | 1030 | 122 | |
XM_011539011.1 | XP_011537313.1 | 7 - 8 | 975 | 122 | |
XM_017020240.1 | XP_016875729.1 | 7 - 8 | 1057 | 122 | |
XM_017020241.1 | XP_016875730.1 | 7 - 8 | 1389 | 122 | |
XM_017020242.1 | XP_016875731.1 | 7 - 8 | 1044 | 122 | |
GenBank mRNA | AB018282.1 | - | 9 - 10 | 1109 | 122 |
AF069512.1 | - | 7 - 8 | 866 | 122 | |
AF107099.1 | - | 7 - 8 | 865 | 122 | |
AK128321.1 | - | 8 - 9 | 1529 | 122 | |
AK295315.1 | - | 7 - 8 | 975 | 122 | |
AK299545.1 | - | 4 - 5 | 661 | 122 | |
AL831915.1 | - | 7 - 8 | 1101 | 122 | |
BC025994.1 | - | 7 - 8 | 917 | 122 | |
DQ063579.2 | - | 7 - 8 | 1018 | 122 | |
DQ975204.1 | - | 7 - 8 | 845 | 122 | |
DQ996398.1 | - | 7 - 8 | 1079 | 122 | |
DQ996537.1 | - | 7 - 8 | 845 | 122 |