Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A18AS
Entrez Gene ID
5003
Gene Name
solute carrier family 22 member 18 antisense
Gene Aliases
BWR1B, BWSCR1B, ORCTL2S, SLC22A1LS, p27-BWR1B
Chromosome Location
Chr.11: 2887344 - 2905504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302862.1 NP_001289791.1 2 - 3 612 89
GenBank mRNA BC030237.1 - 2 - 3 168 89

View Full Product Details