The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198819.1 | NP_001185748.1 | 11 - 12 | 1564 | 70 |
NM_138932.2 | NP_620310.1 | 9 - 10 | 1328 | 70 | |
XM_005269718.2 | XP_005269775.1 | 10 - 11 | 1373 | 70 | |
XM_005269720.3 | XP_005269777.1 | 8 - 9 | 1286 | 70 | |
XM_011539729.2 | XP_011538031.1 | 8 - 9 | 1265 | 70 | |
XM_011539730.2 | XP_011538032.1 | 10 - 11 | 1351 | 70 | |
GenBank mRNA | AJ272078.1 | - | 7 - 8 | 1144 | 70 |
AK303801.1 | - | 11 - 12 | 1518 | 70 | |
BC130519.1 | - | 8 - 9 | 1225 | 70 |