The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198818.1 | NP_001185747.1 | 11 - 12 | 1604 | 104 |
NM_001198819.1 | NP_001185748.1 | 12 - 13 | 1771 | 104 | |
NM_001198820.1 | NP_001185749.1 | 11 - 12 | 1654 | 104 | |
NM_014576.3 | NP_055391.2 | 10 - 11 | 1511 | 104 | |
NM_138932.2 | NP_620310.1 | 10 - 11 | 1535 | 104 | |
NM_138933.2 | NP_620311.1 | 10 - 11 | 1606 | 104 | |
XM_005269718.2 | XP_005269775.1 | 11 - 12 | 1580 | 104 | |
XM_005269720.3 | XP_005269777.1 | 9 - 10 | 1493 | 104 | |
XM_011539729.2 | XP_011538031.1 | 9 - 10 | 1472 | 104 | |
XM_011539730.2 | XP_011538032.1 | 11 - 12 | 1558 | 104 | |
XM_017016160.1 | XP_016871649.1 | 9 - 10 | 1469 | 104 | |
GenBank mRNA | AF209192.1 | - | 10 - 11 | 1463 | 104 |
AF271789.1 | - | 8 - 9 | 1323 | 104 | |
AF271790.1 | - | 9 - 10 | 1473 | 104 | |
AJ272078.1 | - | 8 - 9 | 1351 | 104 | |
AJ272079.1 | - | 8 - 9 | 1327 | 104 | |
AK000324.1 | - | 10 - 11 | 1605 | 104 | |
AK291982.1 | - | 11 - 12 | 1556 | 104 | |
AK303801.1 | - | 12 - 13 | 1725 | 104 | |
BC130519.1 | - | 9 - 10 | 1432 | 104 | |
BC144196.1 | - | 10 - 11 | 1551 | 104 |