Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STX16
Entrez Gene ID
8675
Gene Name
syntaxin 16
Gene Aliases
SYN16
Chromosome Location
Chr.20: 58651253 - 58679526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001433.2 NP_001001433.1 8 - 9 1629 137
NM_001134772.2 NP_001128244.1 7 - 8 1617 137
NM_001134773.2 NP_001128245.1 8 - 9 1578 137
NM_001204868.1 NP_001191797.1 9 - 10 991 137
NM_003763.5 NP_003754.2 7 - 8 1566 137
NR_037941.1 7 - 8 1415 137
NR_037942.1 8 - 9 808 137
NR_037943.1 7 - 8 765 137
GenBank mRNA AF008935.1 - 7 - 8 949 137
AF008936.1 - 8 - 9 1012 137
AF008937.1 - 7 - 8 801 137
AF038897.1 - 8 - 9 821 137
AK026970.1 - 8 - 9 760 137
AK296282.1 - 5 - 6 528 137
AK304867.1 - 8 - 9 1238 137
AK316547.1 - 8 - 9 808 137
BC019042.2 - 7 - 8 1021 137
BC073876.1 - 7 - 8 928 137

View Full Product Details