Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC35C2
- Entrez Gene ID
-
51006
- Gene Name
- solute carrier family 35 member C2
- Gene Aliases
-
BA394O2.1, C20orf5, CGI-15, OVCOV1
- Chromosome Location
-
Chr.20: 46349528 - 46364461 on Build GRCh38
View Full Product Details