Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
Chromosome Location
Chr.20: 46349528 - 46364461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281459.1 NP_001268388.1 5 - 6 684 79
NM_173073.3 NP_775096.1 5 - 6 755 79
XM_017027861.1 XP_016883350.1 5 - 6 917 79
GenBank mRNA AK300798.1 - 5 - 6 616 79
BC014191.2 - 5 - 6 699 79
BI831689.1 - 6 - 7 628 79

View Full Product Details