Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP7
Entrez Gene ID
655
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP-1
Chromosome Location
Chr.20: 57168753 - 57266651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001719.2 NP_001710.1 6 - 7 1679 61
GenBank mRNA AK094784.1 - 2 - 3 419 61
AK291186.1 - 6 - 7 1255 61
AK312419.1 - 6 - 7 1252 61
BC008584.1 - 6 - 7 1191 61
EU679366.1 - 6 - 7 1063 61
KJ875974.1 - 3 - 4 274 61
M60316.1 - 6 - 7 1246 61
X51801.1 - 6 - 7 1272 61

View Full Product Details