The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313905.1 | NP_001300834.1 | 2 - 3 | 149 | 80 |
NM_001425.2 | NP_001416.1 | 2 - 3 | 333 | 80 | |
XM_011526605.2 | XP_011524907.1 | 3 - 4 | 264 | 80 | |
GenBank mRNA | AL541088.3 | - | 2 - 3 | 333 | 80 |
BC009718.1 | - | 2 - 3 | 140 | 80 | |
BT007124.1 | - | 1 - 2 | 79 | 80 | |
CR456871.1 | - | 1 - 2 | 79 | 80 | |
CR541955.1 | - | 1 - 2 | 79 | 80 | |
U52101.1 | - | 2 - 3 | 128 | 80 | |
U87947.1 | - | 2 - 3 | 320 | 80 | |
X94771.1 | - | 1 - 2 | 82 | 80 |