The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291602.1 | NP_001278531.1 | 2 - 3 | 228 | 71 |
NM_006519.3 | NP_006510.1 | 3 - 4 | 270 | 71 | |
GenBank mRNA | AK315601.1 | - | 3 - 4 | 227 | 71 |
BC029412.1 | - | 3 - 4 | 223 | 71 | |
BC105588.1 | - | 3 - 4 | 219 | 71 | |
BU675683.1 | - | 2 - 3 | 71 | ||
CR456931.1 | - | 3 - 4 | 192 | 71 | |
D50663.1 | - | 3 - 4 | 204 | 71 | |
EU862237.1 | - | 3 - 4 | 196 | 71 | |
HY017024.1 | - | 2 - 3 | 181 | 71 | |
HY171700.1 | - | 3 - 4 | 273 | 71 | |
U56255.1 | - | 3 - 4 | 192 | 71 |