The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012393.2 | NP_001012393.1 | 3 - 4 | 667 | 72 |
NM_001319103.1 | NP_001306032.1 | 2 - 3 | 945 | 72 | |
NM_001319104.1 | NP_001306033.1 | 2 - 3 | 582 | 72 | |
NM_001319105.1 | NP_001306034.1 | 3 - 4 | 491 | 72 | |
NM_001319106.1 | NP_001306035.1 | 2 - 3 | 945 | 72 | |
NM_002545.4 | NP_002536.1 | 2 - 3 | 945 | 72 | |
XM_006718846.2 | XP_006718909.1 | 3 - 4 | 893 | 72 | |
XM_011542856.2 | XP_011541158.1 | 4 - 5 | 1203 | 72 | |
GenBank mRNA | AK289695.1 | - | 2 - 3 | 573 | 72 |
AK299528.1 | - | 4 - 5 | 340 | 72 | |
AK299908.1 | - | 3 - 4 | 454 | 72 | |
AK314077.1 | - | 2 - 3 | 945 | 72 | |
AK315859.1 | - | 2 - 3 | 272 | 72 | |
AK316273.1 | - | 2 - 3 | 369 | 72 | |
AK316491.1 | - | 3 - 4 | 491 | 72 | |
BC074742.2 | - | 2 - 3 | 427 | 72 | |
BC074773.2 | - | 2 - 3 | 427 | 72 | |
BC117254.1 | - | 2 - 3 | 423 | 72 | |
BC126251.1 | - | 2 - 3 | 423 | 72 | |
BC143945.1 | - | 2 - 3 | 423 | 72 | |
BC143946.1 | - | 2 - 3 | 423 | 72 | |
BX537377.1 | - | 3 - 4 | 515 | 72 | |
EU562295.1 | - | 2 - 3 | 979 | 72 | |
EU562296.1 | - | 3 - 4 | 457 | 72 | |
EU562297.1 | - | 3 - 4 | 394 | 72 | |
EU562298.1 | - | 2 - 3 | 372 | 72 | |
EU562299.1 | - | 2 - 3 | 284 | 72 | |
EU562300.1 | - | 1 - 2 | 208 | 72 | |
L34774.1 | - | 2 - 3 | 450 | 72 |