The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042610.2 | NP_001036075.1 | 1 - 2 | 247 | 65 |
GenBank mRNA | AK022644.1 | - | 1 - 2 | 73 | 65 |
AL539365.3 | - | 1 - 2 | 252 | 64 | |
BI916419.1 | - | 1 - 2 | 239 | 65 | |
BM747302.1 | - | 1 - 2 | 221 | 65 | |
HY166977.1 | - | 1 - 2 | 228 | 65 |