The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282424.2 | NP_001269353.1 | 9 - 10 | 1281 | 81 |
NM_144670.5 | NP_653271.2 | 20 - 21 | 2766 | 81 | |
XM_011520566.2 | XP_011518868.1 | 20 - 21 | 2723 | 81 | |
XM_011520567.2 | XP_011518869.1 | 20 - 21 | 2723 | 81 | |
XM_017018868.1 | XP_016874357.1 | 20 - 21 | 2720 | 81 | |
XM_017018869.1 | XP_016874358.1 | 20 - 21 | 2720 | 81 | |
XM_017018870.1 | XP_016874359.1 | 20 - 21 | 2684 | 81 | |
GenBank mRNA | AK123591.1 | - | 3 - 4 | 419 | 81 |
AK123592.1 | - | 9 - 10 | 1236 | 81 | |
AK123601.1 | - | 17 - 18 | 2143 | 81 | |
AK290355.1 | - | 20 - 21 | 2619 | 81 | |
AK302555.1 | - | 9 - 10 | 1281 | 81 | |
AL832139.1 | - | 20 - 21 | 2617 | 81 | |
AL833479.1 | - | 18 - 19 | 2282 | 81 | |
BX647674.1 | - | 2 - 3 | 221 | 81 | |
BX648231.1 | - | 21 - 22 | 2618 | 81 |