Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
Chromosome Location
Chr.12: 8822472 - 8887202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282424.2 NP_001269353.1 11 - 12 1462 66
NM_144670.5 NP_653271.2 22 - 23 2947 66
XM_011520566.2 XP_011518868.1 22 - 23 2904 66
XM_011520567.2 XP_011518869.1 22 - 23 2904 66
XM_017018868.1 XP_016874357.1 22 - 23 2901 66
XM_017018869.1 XP_016874358.1 22 - 23 2901 66
XM_017018870.1 XP_016874359.1 22 - 23 2865 66
GenBank mRNA AK123591.1 - 5 - 6 600 66
AK123592.1 - 11 - 12 1417 66
AK123601.1 - 19 - 20 2324 66
AK290355.1 - 22 - 23 2800 66
AK302555.1 - 11 - 12 1462 66
AL832139.1 - 22 - 23 2798 66
AL833479.1 - 20 - 21 2463 66
BX647674.1 - 4 - 5 402 66
BX648231.1 - 23 - 24 2799 66

View Full Product Details