Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
Chromosome Location
Chr.12: 8822472 - 8887202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282424.2 NP_001269353.1 14 - 15 1798 97
NM_144670.5 NP_653271.2 25 - 26 3283 97
XM_011520566.2 XP_011518868.1 25 - 26 3240 97
XM_011520567.2 XP_011518869.1 25 - 26 3240 97
XM_017018868.1 XP_016874357.1 25 - 26 3237 97
XM_017018869.1 XP_016874358.1 25 - 26 3237 97
XM_017018870.1 XP_016874359.1 25 - 26 3201 97
GenBank mRNA AK096448.1 - 3 - 4 272 97
AK123591.1 - 8 - 9 936 97
AK123592.1 - 14 - 15 1753 97
AK123601.1 - 22 - 23 2660 97
AK290355.1 - 25 - 26 3136 97
AK302555.1 - 14 - 15 1798 97
AL832139.1 - 25 - 26 3134 97
AL832750.1 - 2 - 3 102 97
AL833479.1 - 23 - 24 2799 97
BX647674.1 - 7 - 8 738 97
BX648231.1 - 26 - 27 3135 97

View Full Product Details