The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130455.1 | NP_001123927.1 | 49 - 50 | 5668 | 152 |
NM_001130976.1 | NP_001124448.1 | 48 - 49 | 5858 | 152 | |
NM_001130977.1 | NP_001124449.1 | 49 - 50 | 5921 | 152 | |
NM_001130978.1 | NP_001124450.1 | 50 - 51 | 5963 | 152 | |
NM_001130979.1 | NP_001124451.1 | 50 - 51 | 5993 | 152 | |
NM_001130980.1 | NP_001124452.1 | 49 - 50 | 5951 | 152 | |
NM_001130981.1 | NP_001124453.1 | 50 - 51 | 6014 | 152 | |
NM_001130982.1 | NP_001124454.1 | 50 - 51 | 5761 | 152 | |
NM_001130983.1 | NP_001124455.1 | 50 - 51 | 5731 | 152 | |
NM_001130984.1 | NP_001124456.1 | 49 - 50 | 5689 | 152 | |
NM_001130985.1 | NP_001124457.1 | 49 - 50 | 5719 | 152 | |
NM_001130986.1 | NP_001124458.1 | 48 - 49 | 5626 | 152 | |
NM_001130987.1 | NP_001124459.1 | 50 - 51 | 5782 | 152 | |
NM_003494.3 | NP_003485.1 | 49 - 50 | 5900 | 152 | |
XM_005264584.4 | XP_005264641.1 | 51 - 52 | 5839 | 152 | |
XM_005264585.4 | XP_005264642.1 | 51 - 52 | 6524 | 152 | |
GenBank mRNA | AF075575.1 | - | 49 - 50 | 5897 | 152 |
AK074104.1 | - | 35 - 36 | 4170 | 152 | |
AK074649.1 | - | 9 - 10 | 1037 | 152 | |
AK295004.1 | - | 20 - 21 | 2357 | 152 | |
AK303364.1 | - | 16 - 17 | 1971 | 152 | |
BC171803.1 | - | 53 - 54 | 6015 | 152 | |
DQ267935.1 | - | 49 - 50 | 5668 | 152 | |
EU515155.1 | - | 50 - 51 | 5894 | 152 | |
EU515156.1 | - | 48 - 49 | 5759 | 152 | |
EU515157.1 | - | 50 - 51 | 5864 | 152 | |
EU515158.1 | - | 49 - 50 | 5852 | 152 | |
EU515159.1 | - | 49 - 50 | 5822 | 152 | |
EU515160.1 | - | 50 - 51 | 5915 | 152 | |
EU515161.1 | - | 50 - 51 | 5761 | 152 | |
EU515162.1 | - | 48 - 49 | 5626 | 152 | |
EU515163.1 | - | 50 - 51 | 5731 | 152 | |
EU515164.1 | - | 49 - 50 | 5719 | 152 | |
EU515165.1 | - | 49 - 50 | 5689 | 152 | |
EU515166.1 | - | 50 - 51 | 5782 | 152 |