Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC11
Entrez Gene ID
51061
Gene Name
thioredoxin domain containing 11
Gene Aliases
EFP1
Chromosome Location
Chr.16: 11679080 - 11742878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303447.1 NP_001290376.1 1 - 2 402 61
NM_001324022.1 NP_001310951.1 1 - 2 402 61
NM_001324024.1 NP_001310953.1 1 - 2 402 61
NM_001324025.1 NP_001310954.1 1 - 2 402 61
NM_015914.6 NP_056998.4 1 - 2 402 61
NR_136671.1 1 - 2 402 61
NR_136672.1 1 - 2 402 61
NR_136673.1 1 - 2 402 61
NR_136674.1 1 - 2 402 61
XM_011522515.2 XP_011520817.1 1 - 2 359 61
XM_011522516.2 XP_011520818.1 1 - 2 249 61
XM_017023268.1 XP_016878757.1 1 - 2 359 61
GenBank mRNA AK027646.1 - 1 - 2 316 61
BC002856.1 - 1 - 2 371 61
BC018635.2 - 1 - 2 372 61
BX648407.1 - 2 - 3 109 61
CN347909.1 - 1 - 2 402 61

View Full Product Details