Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
Chromosome Location
Chr.13: 50909678 - 50970462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142279.2 NP_001135751.1 2 - 3 533 142
NM_024570.3 NP_078846.2 2 - 3 533 142
XM_011535229.1 XP_011533531.1 2 - 3 455 142
XM_011535230.2 XP_011533532.1 2 - 3 455 142
XM_011535231.2 XP_011533533.1 2 - 3 455 142
XM_011535233.2 XP_011533535.1 2 - 3 203 142
XM_017020747.1 XP_016876236.1 2 - 3 455 142
GenBank mRNA AK223340.1 - 2 - 3 420 142
BC001397.2 - 2 - 3 243 142
BC005088.1 - 2 - 3 243 142
BC007332.2 - 2 - 3 243 142
BC010174.2 - 2 - 3 243 142
BC036744.1 - 2 - 3 404 142
BX382357.2 - 2 - 3 411 142
DB221997.1 - 2 - 3 455 142
DT216141.1 - 2 - 3 455 142

View Full Product Details