Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
Chromosome Location
Chr.13: 50909678 - 50970462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142279.2 NP_001135751.1 3 - 4 641 147
NM_024570.3 NP_078846.2 3 - 4 641 147
XM_006719867.3 XP_006719930.1 3 - 4 330 147
XM_011535229.1 XP_011533531.1 3 - 4 563 147
XM_011535230.2 XP_011533532.1 3 - 4 563 147
XM_011535231.2 XP_011533533.1 3 - 4 563 147
XM_011535233.2 XP_011533535.1 3 - 4 311 147
XM_017020747.1 XP_016876236.1 3 - 4 563 147
GenBank mRNA AK021774.1 - 3 - 4 528 147
AK223340.1 - 3 - 4 528 147
AY764036.1 - 3 - 4 549 147
BC001397.2 - 3 - 4 351 147
BC005088.1 - 3 - 4 351 147
BC007332.2 - 3 - 4 351 147
BC010174.2 - 3 - 4 351 147
BC036744.1 - 3 - 4 512 147
DT216141.1 - 3 - 4 563 147

View Full Product Details