Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
Chromosome Location
Chr.13: 50909678 - 50970462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142279.2 NP_001135751.1 8 - 9 1097 108
NM_024570.3 NP_078846.2 8 - 9 1097 108
XM_006719867.3 XP_006719930.1 8 - 9 786 108
XM_011535229.1 XP_011533531.1 8 - 9 1019 108
XM_011535230.2 XP_011533532.1 8 - 9 1019 108
XM_011535231.2 XP_011533533.1 8 - 9 1019 108
XM_011535233.2 XP_011533535.1 9 - 10 972 108
XM_017020747.1 XP_016876236.1 8 - 9 1019 108
GenBank mRNA AK021774.1 - 8 - 9 984 108
AK124228.1 - 3 - 4 2763 108
AK223340.1 - 8 - 9 984 108
AY764036.1 - 8 - 9 1005 108
BC001397.2 - 8 - 9 807 108
BC005088.1 - 8 - 9 807 108
BC007332.2 - 8 - 9 807 108
BC010174.2 - 8 - 9 807 108
BC036744.1 - 8 - 9 968 108
BX328027.1 - 5 - 6 424 108
BX330672.1 - 2 - 3 108
CA309004.1 - 2 - 3 108

View Full Product Details