The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002044.3 | NP_002035.1 | 2 - 3 | 440 | 147 |
XM_005254279.3 | XP_005254336.1 | 2 - 3 | 229 | 147 | |
XM_006720461.3 | XP_006720524.1 | 2 - 3 | 229 | 147 | |
XM_006720463.3 | XP_006720526.1 | 2 - 3 | 209 | 147 | |
XM_006720464.3 | XP_006720527.1 | 2 - 3 | 209 | 147 | |
GenBank mRNA | BC005141.2 | - | 2 - 3 | 208 | 147 |
BI766538.1 | - | 2 - 3 | 230 | 147 | |
BT006901.1 | - | 2 - 3 | 133 | 147 | |
DB457948.1 | - | 2 - 3 | 440 | 147 | |
DC298656.1 | - | 2 - 3 | 430 | 147 | |
M84443.1 | - | 2 - 3 | 153 | 147 |