The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001556.2 | NP_001001556.1 | 5 - 6 | 709 | 62 |
NM_001289030.1 | NP_001275959.1 | 6 - 7 | 941 | 62 | |
NM_001289031.1 | NP_001275960.1 | 5 - 6 | 794 | 62 | |
NM_002044.3 | NP_002035.1 | 5 - 6 | 811 | 62 | |
XM_005254279.3 | XP_005254336.1 | 5 - 6 | 600 | 62 | |
XM_005254280.3 | XP_005254337.1 | 5 - 6 | 691 | 62 | |
XM_006720461.3 | XP_006720524.1 | 5 - 6 | 600 | 62 | |
XM_006720462.3 | XP_006720525.1 | 5 - 6 | 581 | 62 | |
XM_011521441.2 | XP_011519743.1 | 6 - 7 | 804 | 62 | |
XM_017022064.1 | XP_016877553.1 | 3 - 4 | 657 | 62 | |
XM_017022065.1 | XP_016877554.1 | 6 - 7 | 856 | 62 | |
XM_017022066.1 | XP_016877555.1 | 6 - 7 | 877 | 62 | |
GenBank mRNA | AF461816.1 | - | 5 - 6 | 689 | 62 |
AK098049.1 | - | 6 - 7 | 665 | 62 | |
AK301361.1 | - | 5 - 6 | 564 | 62 | |
AK316440.1 | - | 6 - 7 | 675 | 62 | |
AL832745.1 | - | 5 - 6 | 688 | 62 | |
BC005141.2 | - | 5 - 6 | 579 | 62 | |
BQ921903.1 | - | 5 - 6 | 522 | 62 | |
BT006901.1 | - | 5 - 6 | 504 | 62 | |
EB386877.1 | - | 3 - 4 | 340 | 62 | |
M84443.1 | - | 5 - 6 | 524 | 62 |