The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032802.3 | NP_116191.2 | 9 - 10 | 1190 | 113 |
XM_005254722.3 | XP_005254779.1 | 9 - 10 | 1189 | 113 | |
XM_017022680.1 | XP_016878169.1 | 9 - 10 | 1189 | 113 | |
XM_017022681.1 | XP_016878170.1 | 9 - 10 | 1189 | 113 | |
GenBank mRNA | AJ345028.1 | - | 9 - 10 | 1168 | 113 |
AJ420896.1 | - | 7 - 8 | 741 | 113 | |
AK027446.1 | - | 7 - 8 | 742 | 113 | |
AK075454.1 | - | 9 - 10 | 1179 | 113 | |
AK315651.1 | - | 9 - 10 | 1157 | 113 | |
AY169314.1 | - | 9 - 10 | 1186 | 113 | |
BC025740.1 | - | 9 - 10 | 1153 | 113 |