The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009999.2 | NP_001009999.1 | 1 - 2 | 496 | 95 |
NM_015013.3 | NP_055828.2 | 1 - 2 | 496 | 95 | |
XM_005245786.2 | XP_005245843.1 | 1 - 2 | 501 | 95 | |
XM_006710472.3 | XP_006710535.1 | 1 - 2 | 501 | 95 | |
XM_006710473.2 | XP_006710536.1 | 1 - 2 | 501 | 95 | |
XM_006710474.3 | XP_006710537.1 | 1 - 2 | 501 | 95 | |
XM_017000716.1 | XP_016856205.1 | 1 - 2 | 501 | 95 | |
XM_017000717.1 | XP_016856206.1 | 1 - 2 | 501 | 95 | |
XM_017000719.1 | XP_016856208.1 | 1 - 2 | 130 | 95 | |
XM_017000720.1 | XP_016856209.1 | 1 - 2 | 160 | 95 | |
GenBank mRNA | AB011173.1 | - | 1 - 2 | 449 | 95 |
BC040194.2 | - | 1 - 2 | 451 | 95 | |
BC048134.1 | - | 1 - 2 | 417 | 95 | |
BG717076.1 | - | 1 - 2 | 503 | 95 |