Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM1A
Entrez Gene ID
23028
Gene Name
lysine demethylase 1A
Gene Aliases
AOF2, BHC110, CPRF, KDM1, LSD1
Chromosome Location
Chr.1: 23019443 - 23083691 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009999.2 NP_001009999.1 8 - 9 1214 94
NM_015013.3 NP_055828.2 7 - 8 1154 94
XM_005245786.2 XP_005245843.1 8 - 9 1219 94
XM_006710472.3 XP_006710535.1 7 - 8 1159 94
XM_006710473.2 XP_006710536.1 8 - 9 1219 94
XM_006710474.3 XP_006710537.1 8 - 9 1219 94
XM_017000716.1 XP_016856205.1 8 - 9 1219 94
XM_017000717.1 XP_016856206.1 7 - 8 1159 94
XM_017000718.1 XP_016856207.1 6 - 7 625 94
XM_017000719.1 XP_016856208.1 7 - 8 769 94
XM_017000720.1 XP_016856209.1 6 - 7 739 94
GenBank mRNA AB011173.1 - 7 - 8 1107 94
AK290328.1 - 7 - 8 680 94
AL833812.1 - 3 - 4 276 94
BC016639.1 - 5 - 6 393 94
BC040194.2 - 8 - 9 1169 94
BC048134.1 - 7 - 8 1075 94

View Full Product Details