The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256545.1 | NP_001243474.1 | 17 - 18 | 2543 | 73 |
NM_032446.2 | NP_115822.1 | 18 - 19 | 2594 | 73 | |
XM_011543694.1 | XP_011541996.1 | 17 - 18 | 2306 | 73 | |
XM_017009987.1 | XP_016865476.1 | 19 - 20 | 2839 | 73 | |
XM_017009988.1 | XP_016865477.1 | 10 - 11 | 1612 | 73 | |
GenBank mRNA | AB058676.1 | - | 18 - 19 | 2435 | 73 |
BC152478.1 | - | 18 - 19 | 2435 | 73 | |
CR749437.1 | - | 17 - 18 | 2477 | 73 |