The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280558.1 | NP_001267487.1 | 5 - 6 | 734 | 99 |
NM_006032.3 | NP_006023.1 | 4 - 5 | 722 | 99 | |
XM_005268216.1 | XP_005268273.1 | 5 - 6 | 678 | 99 | |
XM_005268217.1 | XP_005268274.1 | 3 - 4 | 372 | 99 | |
GenBank mRNA | AB009288.1 | - | 4 - 5 | 522 | 99 |
AK222675.1 | - | 4 - 5 | 482 | 99 | |
AK293655.1 | - | 5 - 6 | 734 | 99 | |
AK314182.1 | - | 4 - 5 | 535 | 99 | |
BC018046.1 | - | 4 - 5 | 528 | 99 |