Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPNE6
Entrez Gene ID
9362
Gene Name
copine 6
Gene Aliases
-
Chromosome Location
Chr.14: 24070837 - 24078100 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001280558.1 NP_001267487.1 10 - 11 1177 72
NM_006032.3 NP_006023.1 9 - 10 1165 72
XM_005268216.1 XP_005268273.1 10 - 11 1121 72
XM_005268217.1 XP_005268274.1 8 - 9 815 72
GenBank mRNA AB009288.1 - 9 - 10 965 72
AK125195.1 - 8 - 9 1727 72
AK222675.1 - 9 - 10 925 72
AK293655.1 - 10 - 11 1177 72
AK314182.1 - 9 - 10 978 72
BC018046.1 - 9 - 10 971 72

View Full Product Details