The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001330.2 | NP_001001330.1 | 5 - 6 | 598 | 138 |
GenBank mRNA | BC010040.2 | - | 5 - 6 | 548 | 138 |
BC057832.1 | - | 5 - 6 | 553 | 138 | |
BC068557.1 | - | 5 - 6 | 598 | 138 | |
KJ534927.1 | - | 5 - 6 | 415 | 138 | |
KJ535031.1 | - | 4 - 5 | 342 | 138 |