The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199779.1 | NP_001186708.1 | 2 - 3 | 260 | 109 |
NM_002794.4 | NP_002785.1 | 2 - 3 | 628 | 109 | |
GenBank mRNA | AB209470.1 | - | 2 - 3 | 314 | 109 |
AK296956.1 | - | 2 - 3 | 235 | 109 | |
AV762678.2 | - | 2 - 3 | 322 | 105 | |
BC000268.1 | - | 2 - 3 | 274 | 109 | |
BC101836.1 | - | 2 - 3 | 260 | 109 | |
BC105126.1 | - | 2 - 3 | 260 | 109 | |
BC107901.1 | - | 2 - 3 | 275 | 109 | |
BM849239.1 | - | 2 - 3 | 260 | 109 | |
BT007137.1 | - | 2 - 3 | 216 | 109 | |
CR456862.1 | - | 2 - 3 | 216 | 109 | |
D26599.1 | - | 2 - 3 | 245 | 109 |