The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033549.4 | NP_291027.3 | 4 - 5 | 1894 | 58 |
NM_201627.2 | NP_963921.1 | 4 - 5 | 1894 | 58 | |
XM_006714929.2 | XP_006714992.1 | 4 - 5 | 1892 | 58 | |
XM_006714930.2 | XP_006714993.1 | 4 - 5 | 1892 | 58 | |
XM_006714931.3 | XP_006714994.1 | 4 - 5 | 1892 | 58 | |
XM_006714932.3 | XP_006714995.1 | 4 - 5 | 1892 | 58 | |
GenBank mRNA | AB063180.1 | - | 4 - 5 | 920 | 58 |
AB100366.1 | - | 4 - 5 | 1157 | 58 | |
AB100367.1 | - | 4 - 5 | 1157 | 58 | |
AK027601.1 | - | 4 - 5 | 1867 | 58 | |
AL832960.1 | - | 4 - 4 | 258 | ||
AX775789.1 | - | 4 - 5 | 633 | 58 | |
AX775791.1 | - | 4 - 5 | 633 | 58 | |
AX775793.1 | - | 4 - 5 | 1867 | 58 | |
AX775797.1 | - | 4 - 5 | 925 | 58 | |
AX775799.1 | - | 4 - 5 | 1867 | 58 | |
AX775801.1 | - | 4 - 4 | 258 | ||
AX775803.1 | - | 4 - 4 | 258 | ||
BC018765.1 | - | 4 - 5 | 925 | 58 | |
BC090953.1 | - | 4 - 5 | 1437 | 58 |