Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKAP1
Entrez Gene ID
8631
Gene Name
src kinase associated phosphoprotein 1
Gene Aliases
HEL-S-81p, SCAP1, SKAP55
Chromosome Location
Chr.17: 48133438 - 48430291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003726.3 NP_003717.3 10 - 11 993 136
XM_017025257.1 XP_016880746.1 9 - 10 915 136
XM_017025258.1 XP_016880747.1 7 - 8 1122 136
XM_017025259.1 XP_016880748.1 11 - 12 1130 136
XM_017025260.1 XP_016880749.1 7 - 8 695 136
GenBank mRNA AK290818.1 - 10 - 11 980 136
Y11215.1 - 10 - 11 951 136

View Full Product Details