Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKAP1
Entrez Gene ID
8631
Gene Name
src kinase associated phosphoprotein 1
Gene Aliases
HEL-S-81p, SCAP1, SKAP55
Chromosome Location
Chr.17: 48133438 - 48430291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075099.1 NP_001068567.1 3 - 4 294 91
NM_003726.3 NP_003717.3 3 - 4 294 91
XM_005257755.3 XP_005257812.1 3 - 4 353 91
XM_017025259.1 XP_016880748.1 3 - 4 312 91
GenBank mRNA AK290818.1 - 3 - 4 281 91
BC047870.1 - 3 - 4 266 91
DR007155.1 - 3 - 4 299 91
EU668346.1 - 3 - 4 266 91
Y11215.1 - 3 - 4 252 91

View Full Product Details