Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UIMC1
- Entrez Gene ID
-
51720
- Gene Name
- ubiquitin interaction motif containing 1
- Gene Aliases
-
RAP80, X2HRIP110
- Chromosome Location
-
Chr.5: 176905005 - 177022640 on Build GRCh38
View Full Product Details