Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EYA1
- Entrez Gene ID
-
2138
- Gene Name
- EYA transcriptional coactivator and phosphatase 1
- Gene Aliases
-
BOP, BOR, BOS1, OFC1
- Chromosome Location
-
Chr.8: 71197433 - 71548130 on Build GRCh38
View Full Product Details