The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303245.1 | NP_001290174.1 | 14 - 15 | 1522 | 107 |
NM_001303246.1 | NP_001290175.1 | 14 - 15 | 1522 | 107 | |
NM_006506.3 | NP_006497.2 | 14 - 15 | 1522 | 107 | |
XM_011513059.2 | XP_011511361.1 | 15 - 16 | 2671 | 107 | |
XM_017006968.1 | XP_016862457.1 | 15 - 16 | 1655 | 107 | |
XM_017006969.1 | XP_016862458.1 | 15 - 16 | 1654 | 107 | |
XM_017006970.1 | XP_016862459.1 | 15 - 16 | 1661 | 107 | |
XM_017006971.1 | XP_016862460.1 | 15 - 16 | 1663 | 107 | |
XM_017006972.1 | XP_016862461.1 | 13 - 14 | 1386 | 107 | |
GenBank mRNA | AF115573.1 | - | 14 - 15 | 1485 | 107 |
AK292016.1 | - | 14 - 15 | 1520 | 107 | |
AK301015.1 | - | 15 - 16 | 1461 | 107 | |
D82880.1 | - | 14 - 15 | 1485 | 107 |