Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A27
Entrez Gene ID
9481
Gene Name
solute carrier family 25 member 27
Gene Aliases
UCP4
Chromosome Location
Chr.6: 46652915 - 46678190 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204051.1 NP_001190980.1 4 - 5 782 108
NM_001204052.1 NP_001190981.1 4 - 5 782 108
NM_004277.4 NP_004268.3 4 - 5 782 108
XM_005249483.2 XP_005249540.1 4 - 5 752 108
XM_017011512.1 XP_016867001.1 3 - 4 631 108
GenBank mRNA AF110532.1 - 4 - 5 504 108
AK090871.1 - 2 - 3 1111 108
AK291427.1 - 4 - 5 712 108
AK295233.1 - 4 - 5 765 108
AK302906.1 - 5 - 6 822 108
AK315940.1 - 4 - 5 693 108
AY358711.1 - 4 - 5 530 108
BC033091.1 - 4 - 5 755 108

View Full Product Details