Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYNLT1
Entrez Gene ID
6993
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
CW-1, TCTEL1, tctex-1
Chromosome Location
Chr.6: 158636474 - 158644786 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291603.1 NP_001278532.1 1 - 2 108 81
NM_006519.3 NP_006510.1 1 - 2 108 81
GenBank mRNA AK315601.1 - 1 - 2 65 81
BC029412.1 - 1 - 2 61 81
BC105588.1 - 1 - 2 57 81
D50663.1 - 1 - 2 42 81
HY171700.1 - 1 - 2 111 81

View Full Product Details