Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARHGEF9
- Entrez Gene ID
-
23229
- Gene Name
- Cdc42 guanine nucleotide exchange factor 9
- Gene Aliases
-
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
- Chromosome Location
-
Chr.X: 63634967 - 63786025 on Build GRCh38
View Full Product Details