Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANGPTL6
Entrez Gene ID
83854
Gene Name
angiopoietin like 6
Gene Aliases
AGF, ARP5
Chromosome Location
Chr.19: 10092337 - 10106194 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321411.1 NP_001308340.1 4 - 5 1068 81
NM_031917.2 NP_114123.2 4 - 5 1144 81
XM_005260091.3 XP_005260148.1 4 - 5 1311 81
XM_011528347.2 XP_011526649.1 5 - 6 1173 81
XM_011528348.2 XP_011526650.1 5 - 6 1347 81
XM_011528349.2 XP_011526651.1 6 - 7 1806 81
XM_011528350.2 XP_011526652.1 5 - 6 1658 81
XM_017027347.1 XP_016882836.1 5 - 6 1331 81
GenBank mRNA AB054064.1 - 4 - 5 1144 81
AF230330.1 - 4 - 5 1193 81
AY358276.1 - 4 - 5 1069 81
BC142632.1 - 4 - 5 1062 81

View Full Product Details