The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006407.3 | NP_006398.1 | 2 - 3 | 504 | 72 |
GenBank mRNA | AB014765.1 | - | 2 - 3 | 410 | 72 |
AB052638.1 | - | 2 - 3 | 402 | 72 | |
AB097051.1 | - | 2 - 3 | 485 | 72 | |
AF070523.1 | - | 2 - 3 | 510 | 72 | |
AF115965.1 | - | 2 - 3 | 495 | 72 | |
AF125530.1 | - | 2 - 3 | 496 | 72 | |
AF161476.1 | - | 2 - 3 | 485 | 72 | |
AK074612.1 | - | 2 - 3 | 456 | 72 | |
AK223566.1 | - | 2 - 3 | 408 | 72 | |
AK301637.1 | - | 3 - 4 | 418 | 72 | |
AK303148.1 | - | 2 - 3 | 390 | 72 | |
AK312731.1 | - | 2 - 3 | 407 | 72 | |
AY102608.1 | - | 2 - 3 | 395 | 72 | |
BC005143.2 | - | 2 - 3 | 427 | 72 | |
BC020797.1 | - | 2 - 3 | 421 | 72 |